missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane, 99%

Catalog No. AC106840250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106840250 25 g Glass bottle
AC106841000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106840250 Supplier Thermo Scientific Chemicals Supplier No. 106840250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6940-78-9 | C4H8BrCl | 171.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6940-78-9
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.46
MDL Number MFCD00001010
InChI Key NIDSRGCVYOEDFW-UHFFFAOYSA-N
Synonym butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
PubChem CID 81364
IUPAC Name 1-bromo-4-chlorobutane
SMILES C(CCBr)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.4800g/mL
Boiling Point 80.0°C to 82.0°C (30.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4865 to 1.4885
Linear Formula Cl(CH2)4Br
Quantity 25 g
Specific Gravity 1.48
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with organic solvents,soluble in ethanol,ethylether,slightly soluble in carbontetrachloride
Formula Weight 171.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-chlorobutane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-089-3
TSCA TSCA

RUO – Research Use Only