missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane, 99%

Catalog No. AAA1139914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1139914 25 g
AAA1139922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1139914 Supplier Thermo Scientific Chemicals Supplier No. A1139914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6940-78-9 | C4H8BrCl | 171.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6940-78-9
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00001010
InChI Key NIDSRGCVYOEDFW-UHFFFAOYSA-N
Synonym butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
PubChem CID 81364
IUPAC Name 1-bromo-4-chlorobutane
SMILES C(CCBr)CCl

Specifications

Density 1.489
Boiling Point 175°C to 176°C
Flash Point 60°C (140°F)
Odor Aromatic
Refractive Index 1.4875
Quantity 25 g
UN Number UN1993
Beilstein 1098275
Formula Weight 171.47
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-chlorobutane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 230-089-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only