missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-phenylimidazo[1,5-a]pyridine 97.0+%, TCI America™
$285.21 - $285.21
Chemical Identifiers
| CAS | 104202-15-5 |
|---|---|
| Molecular Formula | C13H9BrN2 |
| Molecular Weight (g/mol) | 273.13 |
| MDL Number | MFCD04587511 |
| InChI Key | ULKZZBGVTIZXSA-UHFFFAOYSA-N |
| PubChem CID | 957540 |
| IUPAC Name | 1-bromo-3-phenylimidazo[1,5-a]pyridine |
| SMILES | BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1 |
Chemical Identifiers
| 104202-15-5 | |
| 273.13 | |
| ULKZZBGVTIZXSA-UHFFFAOYSA-N | |
| 1-bromo-3-phenylimidazo[1,5-a]pyridine |
| C13H9BrN2 | |
| MFCD04587511 | |
| 957540 | |
| BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1 |
Specifications
| 104202-15-5 | |
| MFCD04587511 | |
| ULKZZBGVTIZXSA-UHFFFAOYSA-N | |
| 1-bromo-3-phenylimidazo[1,5-a]pyridine | |
| 957540 | |
| ≥97.0% (GC,T) | |
| 1-Bromo-3-phenylimidazo[1,5-a]pyridine |
| C13H9BrN2 | |
| 1 g | |
| BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1 | |
| 273.13 | |
| 273.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No