missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenylimidazo[1,5-a]pyridine 97.0+%, TCI America™

Catalog No. B34381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34381G Supplier TCI America Supplier No. B34381G
Only null left

Chemical Identifiers

CAS 104202-15-5
Molecular Formula C13H9BrN2
Molecular Weight (g/mol) 273.13
MDL Number MFCD04587511
InChI Key ULKZZBGVTIZXSA-UHFFFAOYSA-N
PubChem CID 957540
IUPAC Name 1-bromo-3-phenylimidazo[1,5-a]pyridine
SMILES BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 273.13
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3-phenylimidazo[1,5-a]pyridine