missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-methoxypropane 98.0+%, TCI America™
Supplier: TCI America B34995G
Specifications
| 1-Bromo-3-methoxypropane | |
| Yellow | |
| C4H9BrO | |
| 5 g | |
| 3-Bromopropyl Methyl Ether | |
| COCCCBr | |
| 153.02 | |
| 153.02 | |
| Liquid |
| 36865-41-5 | |
| 132°C | |
| MFCD02258473 | |
| 1993 | |
| CEVMYGZHEJSOHZ-UHFFFAOYSA-N | |
| 1-bromo-3-methoxypropane | |
| 524551 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 36865-41-5 | |
| 153.02 | |
| CEVMYGZHEJSOHZ-UHFFFAOYSA-N | |
| 524551 | |
| COCCCBr |
| C4H9BrO | |
| MFCD02258473 | |
| 3-Bromopropyl Methyl Ether | |
| 1-bromo-3-methoxypropane |
Safety and Handling
TSCA : No