missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methoxypropane 98.0+%, TCI America™

Supplier:  TCI America B349925G

 View more versions of this product

Catalog No. B349925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Bromo-3-methoxypropane
Yellow
C4H9BrO
25 g
3-Bromopropyl Methyl Ether
COCCCBr
153.02
153.02
Liquid
36865-41-5
132°C
MFCD02258473
1993
CEVMYGZHEJSOHZ-UHFFFAOYSA-N
1-bromo-3-methoxypropane
524551
≥98.0% (GC)

Chemical Identifiers

36865-41-5
153.02
CEVMYGZHEJSOHZ-UHFFFAOYSA-N
524551
COCCCBr
C4H9BrO
MFCD02258473
3-Bromopropyl Methyl Ether
1-bromo-3-methoxypropane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions