missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloropropane 99.0+%, TCI America™

Catalog No. B057525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B057525G 25 g
B0575500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B057525G Supplier TCI America Supplier No. B057525G

Chemical Identifiers

CAS 109-70-6
Molecular Formula C3H6BrCl
Molecular Weight (g/mol) 157.435
MDL Number MFCD00000998
InChI Key MFESCIUQSIBMSM-UHFFFAOYSA-N
Synonym trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane
PubChem CID 8006
IUPAC Name 1-bromo-3-chloropropane
SMILES C(CCl)CBr

Specifications

Melting Point -59°C
Color Colorless
Boiling Point 143°C
Quantity 25 g
UN Number 2688
Formula Weight 157.44
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-chloropropane
EINECSNumber (9)-0370&(9)-1247
RTECSNumber TX4113000
TSCA Yes