missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloropropane, 99%

Catalog No. AC106860501
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106860501 50 mL Glass bottle
AC106862500 250 mL Glass bottle
AC106860010 1 L Glass bottle
AC106860025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC106860501 Supplier Thermo Scientific Chemicals Supplier No. 106860501
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-70-6 | C3H6BrCl | 157.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-70-6
Molecular Formula C3H6BrCl
Molecular Weight (g/mol) 157.44
MDL Number MFCD00000998
InChI Key MFESCIUQSIBMSM-UHFFFAOYSA-N
Synonym trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane
PubChem CID 8006
IUPAC Name 1-bromo-3-chloropropane
SMILES C(CCl)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -59.0°C
Density 1.5900g/mL
Boiling Point 142.0°C to 145.0°C
Flash Point 81°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.4870
Linear Formula Cl(CH2)3Br
Quantity 50 mL
Beilstein 01, 109
Specific Gravity 1.59
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,ether and chloroform
Formula Weight 157.44
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-chloropropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to f
EINECSNumber 203-697-1

RUO – Research Use Only