Learn More
1-Bromo-3-chloropropane, 99%
CAS: 109-70-6 | C3H6BrCl | 157.435 g/mol
Supplier: Thermo Scientific Chemicals A113950B
Description
1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.
Solubility
Miscible with alcohols, chlorofom and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing, reducing agents, amines, nitrides, azo/diazo compounds, alkali metals and epoxides.
Specifications
| 1-Bromo-3-chloropropane | |
| -59°C | |
| 142°C to 145°C | |
| C3H6BrCl | |
| MFCD00000998 | |
| UN2688 | |
| trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane | |
| MFESCIUQSIBMSM-UHFFFAOYSA-N | |
| 1-bromo-3-chloropropane | |
| 8006 | |
| 99% |
| 109-70-6 | |
| 1.598 | |
| >140°C (284°F) | |
| 1.4855 | |
| 1000 g | |
| 605278 | |
| Miscible with alcohols,chlorofom and ether. Immiscible with water. | |
| C(CCl)CBr | |
| 157.435 | |
| 157.44 |
Chemical Identifiers
| 109-70-6 | |
| 157.435 | |
| MFESCIUQSIBMSM-UHFFFAOYSA-N | |
| 8006 | |
| C(CCl)CBr |
| C3H6BrCl | |
| MFCD00000998 | |
| trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane | |
| 1-bromo-3-chloropropane |
Safety and Handling
GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P311-P330-P370+P378q-P501c
H227-H302-H331-H335-H341
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1-BROMO-3-CHLOROPROPANE
EINECSNumber : 203-697-1
RTECSNumber : TX4113000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only