Learn More
1-Bromo-3-chloropropane, 99%
Description
1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.
Solubility
Miscible with alcohols, chlorofom and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing, reducing agents, amines, nitrides, azo/diazo compounds, alkali metals and epoxides.
Specifications
Specifications
| Melting Point | -59°C |
| Density | 1.598 |
| Boiling Point | 142°C to 145°C |
| Flash Point | >140°C (284°F) |
| Refractive Index | 1.4855 |
| Quantity | 250 g |
| UN Number | UN2688 |
| Beilstein | 605278 |
| Solubility Information | Miscible with alcohols,chlorofom and ether. Immiscible with water. |
| Formula Weight | 157.44 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H331-H335-H341 |
| Hazard Statement | GHS H Statement H331-H302 Toxic if inhaled. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P311-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1-BROMO-3-CHLOROPROPANE |
| EINECSNumber | 203-697-1 |
| RTECSNumber | TX4113000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only