missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chlorobenzene 99.0+%, TCI America™

Catalog No. b0570100g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
B0570100G 100 g
B057025G 25 g
2 options

Catalog No. B0570100G

Supplier: TCI America B0570100G

Only null left

Chemical Identifiers

CAS 108-37-2
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000568
InChI Key JRGGUPZKKTVKOV-UHFFFAOYSA-N
Synonym 3-bromochlorobenzene, m-bromochlorobenzene, 3-chlorobromobenzene, benzene, 1-bromo-3-chloro, m-chlorobromobenzene, 1-chloro-3-bromobenzene, 3-chlorophenyl bromide, m-chlorophenyl bromide, m-bromophenyl chloride, 3-bromo-1-chlorobenzene
PubChem CID 7928
IUPAC Name 1-bromo-3-chlorobenzene
SMILES ClC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 196°C
Quantity 100 g
Physical Form Clear Liquid at 20°C
Formula Weight 191.45
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1-Bromo-3-chlorobenzene
EINECSNumber (3)-0066
RTECSNumber CY9005000
TSCA Yes