missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-naphthaldehyde 96.0+%, TCI America™

Catalog No. p-7119181
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16471G 1 g
B16475G 5 g
B164725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B16471G Supplier TCI America Supplier No. B16471G
Only null left

Chemical Identifiers

CAS 3378-82-3
Molecular Formula C11H7BrO
Molecular Weight (g/mol) 235.08
MDL Number MFCD00046368
InChI Key CYGUXEZVBLMVRV-UHFFFAOYSA-N
PubChem CID 76909
IUPAC Name 1-bromonaphthalene-2-carbaldehyde
SMILES BrC1=C2C=CC=CC2=CC=C1C=O

Specifications

Melting Point 118°C
Color Yellow
Quantity 1 g
Formula Weight 235.08
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-naphthaldehyde