missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylnaphthalene, 90%, technical

Catalog No. p-75240
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154260250 25 g Glass bottle
AC154261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154260250 Supplier Thermo Scientific Chemicals Supplier No. 154260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2586-62-1 | C11H9Br | 221.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2586-62-1
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00003871
InChI Key CMIMBQIBIZZZHQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo-2-methyl, 2-methyl-1-bromonaphthalene, .beta.-methyl-.alpha.-bromonaphthalene, zlchem 421, 1-bromo-2-methylnaphtalene, acmc-1cq62, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromonaphthalen-2-yl methyl
PubChem CID 75754
IUPAC Name 1-bromo-2-methylnaphthalene
SMILES CC1=CC=C2C=CC=CC2=C1Br

Specifications

CAS Min % 90.0
CAS Max % 100.0
Color Yellow
Density 1.4100g/mL
Boiling Point 296.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.6480 to 1.6500
Linear Formula CH3C10H6Br
Quantity 25 g
Beilstein 05, 568
Specific Gravity 1.41
Formula Weight 221.1
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methylnaphthalene, 90-95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 219-966-1
TSCA TSCA

RUO – Research Use Only