missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloroethane, 98%

Catalog No. AAA1234936
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1234936 500 g
AAA1234922 100 g
AAA123490E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1234936 Supplier Thermo Scientific Chemicals Supplier No. A1234936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-04-0 | C2H4BrCl | 143.408 g/mol

1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 107-04-0
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00000962
InChI Key IBYHHJPAARCAIE-UHFFFAOYSA-N
Synonym ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
PubChem CID 7849
ChEBI CHEBI:19032
IUPAC Name 1-bromo-2-chloroethane
SMILES C(CBr)Cl

Specifications

Melting Point -18°C
Density 1.726
Boiling Point 106°C to 107°C
Flash Point None
Odor Sweet
Refractive Index 1.49
Linear Formula ClCH2CH2Br
Quantity 500 g
UN Number UN2810
Beilstein 605265
Sensitivity Light sensitive
Solubility Information Soluble in water ( 7g/L (20°C). Soluble in ethanol,ethyl ether,chloroform.
Formula Weight 143.42
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloroethane
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335-H341
Hazard Statement GHS H Statement
H301-H350-H312-H332-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-456-0
RTECSNumber KH6500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only