missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chlorobenzene 99.0+%, TCI America™

Catalog No. B0569500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0569500G 500 g
B056925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0569500G Supplier TCI America Supplier No. B0569500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 694-80-4
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.452
MDL Number MFCD00000532
InChI Key QBELEDRHMPMKHP-UHFFFAOYSA-N
Synonym 2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene
PubChem CID 12754
IUPAC Name 1-bromo-2-chlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Br

Specifications

Color Yellow
Quantity 500 g
Physical Form Clear Liquid at 20°C
Boiling Point 204°C
Formula Weight 191.45
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1-Bromo-2-chlorobenzene
EINECSNumber (3)-0066
TSCA Yes