missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-1,1,2-trifluoro-3-butene, 97%

Catalog No. p-7026194
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2205706 5 g
AAB2205714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2205706 Supplier Thermo Scientific Chemicals Supplier No. B2205706
Only null left

CAS: 374-25-4 | C4H3BrClF3 | 223.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374-25-4
Molecular Formula C4H3BrClF3
Molecular Weight (g/mol) 223.42
MDL Number MFCD00039225
InChI Key VULPFOSLGWWARI-UHFFFAOYNA-N
Synonym 1-bromo-2-chloro-1,1,2-trifluoro-3-butene, 4-bromo-3-chloro-3,4,4-trifluorobutene-1, 1-butene, 4-bromo-3-chloro-3,4,4-trifluoro, 4-bromo-3-chloro-3,4,4-trifluorobutene, 4-bromo-3-chloro-3,4,4-trifluoro-1-butene, acmc-1ahj4, 1-butene,4,4-trifluoro, br-cf2-cfcl-ch=ch2, 1-bromo-2-chloro-1,1,2-trifluorobut-3-ene, 1-butene,4-bromo-3-chloro-3,4,4-trifluoro
PubChem CID 136217
IUPAC Name 4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
SMILES FC(F)(Br)C(F)(Cl)C=C

Specifications

Density 1.678
Boiling Point 98°C to 99°C
Refractive Index 1.407
Quantity 5 g
Beilstein 1758700
Formula Weight 223.43
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-chloro-1,1,2-trifluoro-3-butene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only