missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-(2-methoxyethoxy)ethane, tech. 90%, stab. with sodium carbonate
Description
1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible in water.
Notes
Light Sensitive. Store at 4°C. Protect from heat. Store away from strong bases.
1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible in water.
Notes
Light Sensitive. Store at 4°C. Protect from heat. Store away from strong bases.
Specifications
Specifications
| Density | 1.347 |
| Boiling Point | 156°C |
| Flash Point | 73°C (163°F) |
| Refractive Index | 1.455 |
| Quantity | 5 g |
| Beilstein | 2069579 |
| Sensitivity | Light sensitive |
| Solubility Information | Not miscible in water. |
| Formula Weight | 183.05 |
| Percent Purity | 90% |
| Show More |
Safety and Handling
| Hazard Category | H227 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 259-000-6 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only