missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(2-methoxyethoxy)ethane (stabilized with Na2CO3) 90.0+%, TCI America™

Catalog No. p-7123710
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47365G 5 g
B473625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47365G Supplier TCI America Supplier No. B47365G
Only null left

Chemical Identifiers

CAS 54149-17-6
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000238
InChI Key HUXJXNSHCKHFIL-UHFFFAOYSA-N
Synonym 1-bromo-2-2-methoxyethoxy ethane, 1-2-bromoethoxy-2-methoxyethane, 1-bromo-3,6-dioxaheptane, ethane, 1-2-bromoethoxy-2-methoxy, 2-2-methoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-methoxy-ethane, 1-bromo-2-2-methoxy-ethoxy ethane, mpeg2-br, acmc-1alws, 7-bromo-2,5-dioxaheptane
PubChem CID 123532
IUPAC Name 1-bromo-2-(2-methoxyethoxy)ethane
SMILES COCCOCCBr

Specifications

Color Yellow
Boiling Point 156°C
Quantity 5 g
Formula Weight 183.05
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-(2-methoxyethoxy)ethane (stabilized with Na2CO3)
TSCA No
Recommended Storage Refrigerator