missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-1-chloroethane, 98%

Catalog No. AAL0191004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0191004 2 g
AAL0191009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0191004 Supplier Thermo Scientific Chemicals Supplier No. L0191004
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 593-96-4 | C2H4BrCl | 143.408 g/mol

1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

Solubility
Difficult in mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 593-96-4
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00018842
InChI Key QMSVNDSDEZTYAS-UHFFFAOYSA-N
Synonym ch3chclbr, ethane, 1-bromo-1-chloro, bromochloroethane, ethane, bromochloro, bromochloroethan, chlorobromoethane, bromo-chloroethane, 1-bromochoroethane, bromo chloro ethane, acmc-20aoxm
PubChem CID 11654
IUPAC Name 1-bromo-1-chloroethane
SMILES CC(Cl)Br

Specifications

Density 1.667
Boiling Point 82°C to 83°C
Refractive Index 1.466
Quantity 2 g
UN Number UN2810
Beilstein 1730843
Solubility Information Difficult in mix.
Formula Weight 143.42
Percent Purity 98%
Chemical Name or Material 1-Bromo-1-chloroethane
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 209-820-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only