missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-pyrrolidine, 98%, Thermo Scientific Chemicals
$133.64 - $133.64
Chemical Identifiers
| CAS | 86953-79-9 |
|---|---|
| Molecular Formula | C9H17NO2 |
| Molecular Weight (g/mol) | 171.24 |
| MDL Number | MFCD00216581 |
| InChI Key | LPQZERIRKRYGGM-UHFFFAOYSA-N |
| Synonym | 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine |
| PubChem CID | 643455 |
| IUPAC Name | tert-butyl pyrrolidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1747506
|
Thermo Scientific Chemicals
L1747506 |
5 g |
Each for $133.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 86953-79-9 | |
| 171.24 | |
| LPQZERIRKRYGGM-UHFFFAOYSA-N | |
| 643455 | |
| CC(C)(C)OC(=O)N1CCCC1 |
| C9H17NO2 | |
| MFCD00216581 | |
| 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine | |
| tert-butyl pyrrolidine-1-carboxylate |
Specifications
| 86953-79-9 | |
| 130°C (20 mmHg) | |
| C9H17NO2 | |
| MFCD00216581 | |
| 4664750 | |
| LPQZERIRKRYGGM-UHFFFAOYSA-N | |
| tert-butyl pyrrolidine-1-carboxylate | |
| 643455 | |
| 98% |
| 0.977 | |
| 86°C (186°F) | |
| 1.449 | |
| 5 g | |
| 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine | |
| CC(C)(C)OC(=O)N1CCCC1 | |
| 171.24 | |
| 171.24 | |
| 1-Boc-pyrrolidine |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only