missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pyrrolidine-3-carboxylic acid, 99%

CAS: 59378-75-5 | C10H17NO4 | 215.249 g/mol

Supplier:  Thermo Scientific Chemicals H5008003

 View more versions of this product

Catalog No. AAH5008003

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Boc-pyrrolidine-3-carboxylic acid
128°C to 137°C
MFCD02179181
1-boc-pyrrolidine-3-carboxylic acid, 1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-pyrrolidine-3-carboxylic acid, 1-n-boc-beta-proline, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester, 3-carboxy-1-boc-pyrrolidine, 1-tert-butoxy carbonyl pyrrolidine-3-carboxylic acid, 1-n-boc-belta-proline, boc-1-pyrrolidine-3-carboxylic acid, 1-boc-3-pyrrolidinecarboxylic acid
CC(C)(C)OC(=O)N1CCC(C1)C(=O)O
215.249
215.25
59378-75-5
C10H17NO4
1 g
HRMRQBJUFWFQLX-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
2760529
99%

Chemical Identifiers

59378-75-5
215.249
HRMRQBJUFWFQLX-UHFFFAOYSA-N
2760529
CC(C)(C)OC(=O)N1CCC(C1)C(=O)O
C10H17NO4
MFCD02179181
1-boc-pyrrolidine-3-carboxylic acid, 1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-pyrrolidine-3-carboxylic acid, 1-n-boc-beta-proline, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester, 3-carboxy-1-boc-pyrrolidine, 1-tert-butoxy carbonyl pyrrolidine-3-carboxylic acid, 1-n-boc-belta-proline, boc-1-pyrrolidine-3-carboxylic acid, 1-boc-3-pyrrolidinecarboxylic acid
1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only