missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pyrrolidine-3-carboxylic acid, 99%

Catalog No. AAH50080MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50080MD 250 mg
AAH5008003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50080MD Supplier Thermo Scientific Chemicals Supplier No. H50080MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59378-75-5 | C10H17NO4 | 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59378-75-5
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD02179181
InChI Key HRMRQBJUFWFQLX-UHFFFAOYSA-N
Synonym 1-boc-pyrrolidine-3-carboxylic acid, 1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-pyrrolidine-3-carboxylic acid, 1-n-boc-beta-proline, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester, 3-carboxy-1-boc-pyrrolidine, 1-tert-butoxy carbonyl pyrrolidine-3-carboxylic acid, 1-n-boc-belta-proline, boc-1-pyrrolidine-3-carboxylic acid, 1-boc-3-pyrrolidinecarboxylic acid
PubChem CID 2760529
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)O

Specifications

Melting Point 128°C to 137°C
Quantity 250 mg
Formula Weight 215.25
Percent Purity 99%
Chemical Name or Material 1-Boc-pyrrolidine-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only