missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-pyrrolidine-2-methanol, 98%

Catalog No. AAH3278606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3278606 5 g
AAH3278603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3278606 Supplier Thermo Scientific Chemicals Supplier No. H3278606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 170491-63-1 | C10H19NO3 | 201.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170491-63-1
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD01456556
InChI Key BFFLLBPMZCIGRM-UHFFFAOYSA-N
Synonym tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-2-hydroxymethyl pyrrolidine, n-boc-2-pyrrolidinemethanol, boc-dl-prolinol, 2-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 2-hydroxymethyl-1-pyrrolidinecarboxylate, 2-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 2-hydroxymethyl-, 1,1-dimethylethyl ester, r-1-boc-2-hydroxymethyl pyrrolidine, n-boc-dl-prolinol
PubChem CID 550865
IUPAC Name tert-butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 52°C to 54°C
Quantity 5 g
Formula Weight 201.27
Percent Purity 98%
Chemical Name or Material (+/-)-1-Boc-pyrrolidine-2-methanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only