missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-L-nipecotic acid, 97%

Catalog No. p-7034875
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5257506 5 g
AAH5257514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5257506 Supplier Thermo Scientific Chemicals Supplier No. H5257506

CAS: 88495-54-9 | C11H19NO4 | 229.276 g/mol

1-Boc-L-nipecotic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-L-nipecotic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 88495-54-9
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD02179172
InChI Key NXILIHONWRXHFA-QMMMGPOBSA-N
Synonym s-1-boc-piperidine-3-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, s-n-boc-piperidine-3-carboxylic acid, l-1-boc-nipecotic acid, s-boc-nipecotic acid, boc-s-nipecotic acid, boc-s-nip-oh, s-boc-nip, s-n-boc-nipecotic acid, boc-nip-oh
PubChem CID 6951168
IUPAC Name (3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O

Specifications

Melting Point 165°C to 169°C
Quantity 5 g
Optical Rotation +57° (c=1 in Chloroform)
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material 1-Boc-L-nipecotic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only