missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-isonipecotic acid, 98+%

Catalog No. p-7047074
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1752703 1 g
AAL1752706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1752703 Supplier Thermo Scientific Chemicals Supplier No. L1752703

CAS: 84358-13-4 | C11H19NO4 | 229.28 g/mol

It is a pharmaceutical intermediate. he camptothecin ester of isonipecotic acid is installed on a triazine dendrimer intermediate obtained through an iterative, scalable route to ultimately yield cationic and PEGylated targets with activities in cell culture comparable to free drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate. he camptothecin ester of isonipecotic acid is installed on a triazine dendrimer intermediate obtained through an iterative, scalable route to ultimately yield cationic and PEGylated targets with activities in cell culture comparable to free drug.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 84358-13-4
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00076999
InChI Key JWOHBPPVVDQMKB-UHFFFAOYSA-N
Synonym n-boc-isonipecotic acid, 1-tert-butoxycarbonyl piperidine-4-carboxylic acid, n-boc-piperidine-4-carboxylic acid, 1-boc-piperidine-4-carboxylic acid, boc-inp-oh, 1-boc-isonipecotic acid, 1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, boc-isonipecotic acid, 1-boc-4-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-4-carboxylic acid
PubChem CID 392871
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(O)=O

Specifications

Melting Point 149°C to 151°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 229.28
Percent Purity ≥98%
Chemical Name or Material 1-Boc-isonipecotic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only