missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indoline-7-carboxylic acid, 97%

Catalog No. p-7055333
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6388306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6388306 Supplier Thermo Scientific Chemicals Supplier No. H6388306
Only null left

CAS: 143262-20-8 | C14H17NO4 | 263.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143262-20-8
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD04973983
InChI Key SUAMIYWLXFROHE-UHFFFAOYSA-N
Synonym n-boc-indoline-7-carboxylic acid, 1-tert-butoxycarbonyl indoline-7-carboxylic acid, 2,3-dihydro-indole-1,7-dicarboxylic acid 1-tert-butyl ester, 1-tert-butoxycarbonyl-2,3-dihydroindole-7-carboxylic acid, n-boc-indoline-7-carboxylicacid, acmc-1c2ys, 1-tert-butoxycarbonyl-7-indolinecarboxylic acid, 1h-indole-1,7-dicarboxylic acid,2,3-dihydro-,1-1,1-dimethylethyl ester, 11bs-2,6-bis 3,5-bis trifluoromethyl phenyl-4-hydroxy-dinaphtho 2,1-d:1 inverted exclamation marka,2 inverted exclamation marka-f-1,3,2-dioxaphosphepin 4-oxide
PubChem CID 14977845
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-7-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC2=CC=CC(=C21)C(=O)O

Specifications

Melting Point 182°C (decomposition)
Color Yellow
Quantity 5 g
UN Number UN3077
Sensitivity Light sensitive
Formula Weight 263.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-indoline-7-carboxylic acid
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only