missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indole-2-boronic acid, 95%

CAS: 213318-44-6 | C13H16BNO4 | 261.084 g/mol

Supplier:  Thermo Scientific Chemicals L1800906

 View more versions of this product

Catalog No. AAL1800906

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Boc-indole-2-boronic acid
∽85°C (decomposition)
MFCD02093045
8148683
SVIBPSNFXYUOFT-UHFFFAOYSA-N
[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
2773302
95%
213318-44-6
C13H16BNO4
5 g
n-boc-indole-2-boronic acid, 1-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl boronic acid, 1-n-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl-2-boronic acid, 1-tert-butoxycarbonyl indol-2-ylboronic acid, 1-boc-2-indoleboronic acid, n-boc-indol-2-yl boronic acid
B(C1=CC2=CC=CC=C2N1C(=O)OC(C)(C)C)(O)O
261.084
261.08

Chemical Identifiers

213318-44-6
261.084
SVIBPSNFXYUOFT-UHFFFAOYSA-N
2773302
B(C1=CC2=CC=CC=C2N1C(=O)OC(C)(C)C)(O)O
C13H16BNO4
MFCD02093045
n-boc-indole-2-boronic acid, 1-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl boronic acid, 1-n-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl-2-boronic acid, 1-tert-butoxycarbonyl indol-2-ylboronic acid, 1-boc-2-indoleboronic acid, n-boc-indol-2-yl boronic acid
[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only