missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indole-2-boronic acid, 95%

Catalog No. AAL18009MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18009MD 250 mg
AAL1800903 1 g
AAL1800906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL18009MD Supplier Thermo Scientific Chemicals Supplier No. L18009MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 213318-44-6 | C13H16BNO4 | 261.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 213318-44-6
Molecular Formula C13H16BNO4
Molecular Weight (g/mol) 261.084
MDL Number MFCD02093045
InChI Key SVIBPSNFXYUOFT-UHFFFAOYSA-N
Synonym n-boc-indole-2-boronic acid, 1-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl boronic acid, 1-n-boc-indole-2-boronic acid, 1-tert-butoxycarbonyl-1h-indol-2-yl-2-boronic acid, 1-tert-butoxycarbonyl indol-2-ylboronic acid, 1-boc-2-indoleboronic acid, n-boc-indol-2-yl boronic acid
PubChem CID 2773302
IUPAC Name [1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES B(C1=CC2=CC=CC=C2N1C(=O)OC(C)(C)C)(O)O

Specifications

Melting Point ∽85°C (decomposition)
Quantity 250 mg
Beilstein 8148683
Formula Weight 261.08
Percent Purity 95%
Chemical Name or Material 1-Boc-indole-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only