missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-imidazole, 98%

Catalog No. AAL0050606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0050606 5 g
AAL0050614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0050606 Supplier Thermo Scientific Chemicals Supplier No. L0050606

CAS: 49761-82-2 | C8H12N2O2 | 168.196 g/mol

1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N?-carbonyldiimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N′-carbonyldiimidazole.

Solubility
Soluble in hexane, acetone, and DMF.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 49761-82-2
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00014497
InChI Key MTBKGWHHOBJMHJ-UHFFFAOYSA-N
Synonym n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p
PubChem CID 521262
IUPAC Name tert-butyl imidazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=CN=C1

Specifications

Melting Point 44°C to 48°C
Quantity 5 g
Beilstein 607792
Solubility Information Soluble in hexane,acetone,and DMF.
Formula Weight 168.2
Percent Purity 98%
Chemical Name or Material 1-Boc-imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 256-475-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only