missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-hexahydro-1,4-diazepine, 98%

Catalog No. AC399530050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399530050 Supplier Thermo Scientific Chemicals Supplier No. 399530050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112275-50-0 | C10H20N2O2 | 200.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112275-50-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD00276987
InChI Key WDPWEXWMQDRXAL-UHFFFAOYSA-N
Synonym 1-boc-homopiperazine, 1-boc-hexahydro-1,4-diazepine, 1,4 diazepane-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl homopiperazine, 1-boc-1,4-diazepane, n-boc-homopiperazine, boc-hopiz, boc-homopiperazine, n-tert-butoxycarbonyl homopiperazine, tert-butyl 1-homopiperazinecarboxylate
PubChem CID 2756058
IUPAC Name tert-butyl 1,4-diazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCNCC1

Specifications

Boiling Point 92.0°C to 96.0°C (0.5mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Density 1.0290g/mL
Quantity 5 g
Flash Point >110°C
Formula Weight 200.28
Percent Purity 98%
Specific Gravity 1.029
Physical Form Liquid
Chemical Name or Material 1-BOC-Hexahydro-1, 4-diazepine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only