missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-azetidine-3-methanol, 96%, Thermo Scientific Chemicals

Catalog No. AC452090050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC452090050 Supplier Thermo Scientific Chemicals Supplier No. 452090050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142253-56-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD06656141
InChI Key HXRDRJKAEYHOBB-UHFFFAOYSA-N
PubChem CID 10583745
IUPAC Name tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CO)C1

Specifications

Melting Point 50°C to 53°C
Flash Point 117°C
Assay Percent Range 95% min. (GC)
Quantity 5 g
Synonym tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-ylmethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-3-azetidinemethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl3-hydroxymethyl azetidine-1-carboxylate, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester
Formula Weight 187.24
Percent Purity 96%
Chemical Name or Material 1-BOC-azetidine-3-methanol
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only