missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH3435203
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
AAH3435203 1 g
1 options

Catalog No. AAH3435203

Supplier: Thermo Scientific Chemicals H3435203

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 243641-37-4
Molecular Formula C14H20N2O2
Molecular Weight (g/mol) 248.326
MDL Number MFCD06659018
InChI Key CHWMFCCGEWQABK-UHFFFAOYSA-N
Synonym 7-methyl-3,4-dihydro-2h-1,8 naphthyridine-1-carboxylic acid tert-butyl ester, tert-butyl 7-methyl-3,4-dihydro-1,8-naphthyridine-1 2h-carboxylate, 1-boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 1-boc-7-methyl-3,4-dihydro-2h-1,8 naphthyridine, 7-methyl-3,4-dihydro-2h-1,8-naphthyridine-1-carboxylic acid tert-butyl ester, 2-methyl-8-tert-butoxycarbonyl-5,6,7,8-tetrahydro-1,8-naphthyridine, tert-butyl 7-methyl-1,2,3,4-tetrahydropyridino 2,3-b pyridinecarboxylate, tert-butyl 7-methyl-3,4-dihydro-2h-1,8-naphthyridine-1-carboxylate
PubChem CID 17750173
IUPAC Name tert-butyl 7-methyl-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate
SMILES CC1=NC2=C(CCCN2C(=O)OC(C)(C)C)C=C1

Specifications

Melting Point 97°C to 100°C
Quantity 1 g
Formula Weight 248.33
Percent Purity 95%
Chemical Name or Material 1-Boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only