missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-5-methoxyindole-2-boronic acid, 95%

Catalog No. AAH5265403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5265403 1 g
AAH5265406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5265403 Supplier Thermo Scientific Chemicals Supplier No. H5265403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 290331-71-4 | C14H18BNO5 | 291.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 290331-71-4
Molecular Formula C14H18BNO5
Molecular Weight (g/mol) 291.11
MDL Number MFCD04039006
InChI Key PZLVPMBCKHDVKT-UHFFFAOYSA-N
Synonym 1-tert-butoxycarbonyl-5-methoxy-1h-indol-2-ylboronic acid, n-boc-5-methoxy-2-indolylboronic acid, 1-boc-5-methoxyindole-2-boronic acid, 1-tert-butoxycarbonyl-5-methoxy-1h-indol-2-yl boronic acid, n-boc-5-methoxyindole-2-boronic acid, 1-boc-5-methoxy-1h-indole-2-boronic acid, 1-tert-butoxycarbonyl-5-methoxyindol-2-ylboronic acid, pubchem24022, acmc-1cjok
PubChem CID 2794716
IUPAC Name [5-methoxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES COC1=CC=C2N(C(=O)OC(C)(C)C)C(=CC2=C1)B(O)O

Specifications

Melting Point 105°C to 108°C
Quantity 1 g
Formula Weight 291.11
Percent Purity 95%
Chemical Name or Material 1-Boc-5-methoxyindole-2-boronic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only