missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(6-nitro-3-pyridyl)piperazine, 97%

Catalog No. p-7036206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5446503 1 g
AAH5446506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5446503 Supplier Thermo Scientific Chemicals Supplier No. H5446503
Only null left

CAS: 571189-16-7 | C14H20N4O4 | 308.338 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 571189-16-7
Molecular Formula C14H20N4O4
Molecular Weight (g/mol) 308.338
MDL Number MFCD11849291
InChI Key SUWKOEMQNOBJEQ-UHFFFAOYSA-N
Synonym tert-butyl 4-6-nitropyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-nitro-3-pyridyl piperazine, 1-boc-4-6-nitropyridin-3-yl piperazine, 1-piperazinecarboxylic acid, 4-6-nitro-3-pyridinyl-, 1,1-dimethylethyl ester, 4-6-nitro-pyridin-3-yl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-6-nitro-pyridin-3-yl-piperazine, 1-piperazinecarboxylic acid, 4-6-nitro-3-pyridiny, tert-butyl 4-6-nitropyridin-3-yl piperazine-carboxylate, tert-butyl 4-6-nitro-3-pyridyl piperazine-1-carboxylate
PubChem CID 11243758
IUPAC Name tert-butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CN=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 308.33
Percent Purity 97%
Chemical Name or Material 1-Boc-4-(6-nitro-3-pyridyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only