missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(3-hydroxypropyl)piperazine, 97%

Catalog No. p-7052734
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5554706 5 g
AAH5554714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5554706 Supplier Thermo Scientific Chemicals Supplier No. H5554706
Only null left

CAS: 132710-90-8 | C12H24N2O3 | 244.335 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132710-90-8
Molecular Formula C12H24N2O3
Molecular Weight (g/mol) 244.335
MDL Number MFCD06798090
InChI Key LRYRQGKGCIUVON-UHFFFAOYSA-N
Synonym 1-boc-4-3-hydroxypropyl piperazine, tert-butyl 4-3-hydroxypropyl piperazine-1-carboxylate, 4-2-hydroxypropyl-1-piperazinecarboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-3-hydroxypropyl piperazine, 1-piperazinecarboxylicacid, 4-3-hydroxypropyl-, 1,1-dimethylethyl ester, acmc-20dtb2, t-butyl 4-3-hydroxypropyl piperazine-1-carboxylate, 1-3-hydroxypropyl-4-tert-butyloxycarbonylpiperazine
PubChem CID 16217800
IUPAC Name tert-butyl 4-(3-hydroxypropyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCCO

Specifications

Melting Point 73°C to 77°C
Quantity 5 g
Formula Weight 244.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(3-hydroxypropyl)piperazine
Hazard Category H301-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only