missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-hydroxyethyl)piperazine, 97%

Catalog No. aah5599106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5599106 5 g
AAH5599114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5599106 Supplier Thermo Scientific Chemicals Supplier No. H5599106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77279-24-4 | C11H22N2O3 | 230.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77279-24-4
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
MDL Number MFCD00728947
InChI Key VRXIOAYUQIITBU-UHFFFAOYSA-N
Synonym tert-butyl 4-2-hydroxyethyl piperazine-1-carboxylate, 1-boc-4-2-hydroxyethyl piperazine, 1-tert-butyloxy carbonyl-4-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-hydroxyethyl-piperazine, 4-2-hydroxy-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-2-hydroxyethyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-2-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl-1-piperazinecarboxylate, 4-2-hydroxyethyl piperazine-1-carboxylic acid tert-butyl ester
PubChem CID 735736
IUPAC Name tert-butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCO

Specifications

Melting Point 38°C to 42°C
Color Brown to White
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2811
Formula Weight 230.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(2-hydroxyethyl)piperazine
Hazard Category H314
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only