missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-hydroxyethyl)piperazine, 97%

CAS: 77279-24-4 | C11H22N2O3 | 230.308 g/mol

$156.23 - $596.67

Chemical Identifiers

CAS 77279-24-4
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
MDL Number MFCD00728947
InChI Key VRXIOAYUQIITBU-UHFFFAOYSA-N
Synonym tert-butyl 4-2-hydroxyethyl piperazine-1-carboxylate, 1-boc-4-2-hydroxyethyl piperazine, 1-tert-butyloxy carbonyl-4-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-hydroxyethyl-piperazine, 4-2-hydroxy-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-2-hydroxyethyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-2-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl-1-piperazinecarboxylate, 4-2-hydroxyethyl piperazine-1-carboxylic acid tert-butyl ester
PubChem CID 735736
IUPAC Name tert-butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5599106
View Documents
Thermo Scientific Chemicals
H5599106
5 g
Each for $156.23
Only null left
 
AAH5599114
View Documents
Thermo Scientific Chemicals
H5599114
25 g
Each for $596.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

77279-24-4
230.308
VRXIOAYUQIITBU-UHFFFAOYSA-N
735736
CC(C)(C)OC(=O)N1CCN(CC1)CCO
C11H22N2O3
MFCD00728947
tert-butyl 4-2-hydroxyethyl piperazine-1-carboxylate, 1-boc-4-2-hydroxyethyl piperazine, 1-tert-butyloxy carbonyl-4-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-hydroxyethyl-piperazine, 4-2-hydroxy-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-2-hydroxyethyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-2-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl-1-piperazinecarboxylate, 4-2-hydroxyethyl piperazine-1-carboxylic acid tert-butyl ester
tert-butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate

Specifications

77279-24-4
Brown to White
C11H22N2O3
5 g
tert-butyl 4-2-hydroxyethyl piperazine-1-carboxylate, 1-boc-4-2-hydroxyethyl piperazine, 1-tert-butyloxy carbonyl-4-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-hydroxyethyl-piperazine, 4-2-hydroxy-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-2-hydroxyethyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-2-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl-1-piperazinecarboxylate, 4-2-hydroxyethyl piperazine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCN(CC1)CCO
230.308
230.31
Solid
38°C to 42°C
>110°C (230°F)
MFCD00728947
UN2811
VRXIOAYUQIITBU-UHFFFAOYSA-N
tert-butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
735736
97%
1-Boc-4-(2-hydroxyethyl)piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only