missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-chloroethyl)piperazine, 97%

Catalog No. AAH3334906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3334906 5 g
AAH3334903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3334906 Supplier Thermo Scientific Chemicals Supplier No. H3334906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 208167-83-3 | C11H21ClN2O2 | 248.751 g/mol

By the direct condensation of chloroalkyl piperazine, nitrogen mustard benzaldehyde, and pyrrole anticancer activity to bel-7404 liver cancer cells was found.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
By the direct condensation of chloroalkyl piperazine, nitrogen mustard benzaldehyde, and pyrrole anticancer activity to bel-7404 liver cancer cells was found.

Solubility
Slightly Soluble (7.2 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 208167-83-3
Molecular Formula C11H21ClN2O2
Molecular Weight (g/mol) 248.751
MDL Number MFCD08443993
InChI Key MYOWELLYEZMECA-UHFFFAOYSA-N
Synonym tert-butyl 4-2-chloroethyl piperazine-1-carboxylate, tert-butyl 4-2-chloroethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-chloroethyl piperazine, tert-butyl 4-2-chloroethyl piperazinecarboxylate, 1-piperazinecarboxylic acid, 4-2-chloroethyl-, 1,1-dimethylethyl ester, 4-2-chloroethyl piperazine, n1-boc protected, 4-2-chloroethyl piperazine,n1-boc protected, tertbutylchloroethyltetrahydropyrazinecarboxylate, tert-butyl 4-2-chloroethyl tetrahydro-1 2h-pyraz
PubChem CID 22106269
IUPAC Name tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCCl

Specifications

Melting Point 63°C to 65°C
Quantity 5 g
Solubility Information Slightly Soluble (7.2g/L) (25°C).
Formula Weight 248.76
Percent Purity 97%
Chemical Name or Material 1-Boc-4-(2-chloroethyl)piperazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only