missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-4-(2-aminoethyl)piperazine, 95%

Catalog No. p-7077495
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454620050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454620050 Supplier Thermo Scientific Chemicals Supplier No. 454620050

CAS: 192130-34-0 | C11H25N3O2 | 231.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 192130-34-0
Molecular Formula C11H25N3O2
Molecular Weight (g/mol) 231.34
MDL Number MFCD02683049
InChI Key QSYTWBKZNNEKPN-UHFFFAOYSA-P
Synonym tert-butyl 4-2-aminoethyl piperazine-1-carboxylate, 1-boc-4-2-aminoethyl piperazine, 4-n-2-aminoethyl-1-n-boc-piperazine, 4-2-amino-ethyl-1-boc-piperazine, 4-2-amino-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminoethyl-1-tert-butoxycarbonyl piperazine, 1-piperazinecarboxylic acid, 4-2-aminoethyl-, 1,1-dimethylethyl ester, 1-n-boc-4-2-aminoethyl piperazine, 4-2-aminoethyl piperazine-1-carboxylic acid tert-butyl ester, 1-2-aminoethyl-4-tert-butoxycarbonylpiperazine
PubChem CID 1514400
IUPAC Name tert-butyl 4-(2-aminoethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC[NH+](CC[NH3+])CC1

Specifications

Melting Point 36.0°C to 41.0°C
Boiling Point 198.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 229.32
Percent Purity 95%
Chemical Name or Material 1-BOC-4-(2-aminoethyl)piperazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only