missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-pyrrolidinone, 97%

Catalog No. p-7029548
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2702303 1 g
AAH2702309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2702303 Supplier Thermo Scientific Chemicals Supplier No. H2702303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101385-93-7 | C9H15NO3 | 185.223 g/mol

In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 101385-93-7
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD01631194
InChI Key JSOMVCDXPUXKIC-UHFFFAOYSA-N
Synonym n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone
PubChem CID 471360
IUPAC Name tert-butyl 3-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C1

Specifications

Melting Point 34°C to 38°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material 1-Boc-3-pyrrolidinone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only