missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-oxopiperazine, 98%

Catalog No. AAH2782303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2782303 1 g
AAH2782306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2782303 Supplier Thermo Scientific Chemicals Supplier No. H2782303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 76003-29-7 | C9H16N2O3 | 200.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76003-29-7
Molecular Formula C9H16N2O3
Molecular Weight (g/mol) 200.238
MDL Number MFCD02181069
InChI Key FCMLWBBLOASUSO-UHFFFAOYSA-N
Synonym 1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine
PubChem CID 3157178
IUPAC Name tert-butyl 3-oxopiperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNC(=O)C1

Specifications

Melting Point 156°C to 160°C
Quantity 1 g
Formula Weight 200.23
Percent Purity 98%
Chemical Name or Material 1-Boc-3-oxopiperazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only