missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxypiperidine, 97%

Catalog No. AAL1760006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1760006 5 g
AAL1760003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1760006 Supplier Thermo Scientific Chemicals Supplier No. L1760006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85275-45-2 | C10H19NO3 | 201.266 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.

Chemical Identifiers

CAS 85275-45-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD02093938
InChI Key UIJXHKXIOCDSEB-UHFFFAOYSA-N
Synonym 1-boc-3-hydroxypiperidine, 1-n-boc-3-hydroxy-piperidine, 1-boc-3-hydroxy-piperidine, n-boc-3-piperidinol, n-boc-3-hydroxypiperidine, 1-tert-butoxycarbonyl-3-hydroxy piperidine, boc-3-hydroxypiperidine, n-tert-butoxycarbonyl-3-piperidinol, n-boc-3-hydroxy piperidine, 3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 545699
IUPAC Name tert-butyl 3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)O

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Beilstein 4180859
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material 1-Boc-3-hydroxypiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only