missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-bromopyrrolidine, 95%

Catalog No. p-7038167
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5707103 1 g
AAH5707106 5 g
2 options

Catalog No. AAH5707103

Supplier: Thermo Scientific Chemicals H5707103

Only null left

CAS: 939793-16-5 | C9H16BrNO2 | 250.14 g/mol

(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 939793-16-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD08752478
InChI Key QJTKPXFJOXKUEY-UHFFFAOYNA-N
PubChem CID 42614217
IUPAC Name tert-butyl 3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Br)C1

Specifications

Melting Point 45°C to 53°C
Quantity 1 g
UN Number UN3077
Synonym n-boc-3-bromopyrrolidine, 1-boc-3-bromopyrrolidine, 3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, +/--1-boc-3-bromopyrrolidine, acmc-209zg9, ^+-1-boc-3-bromopyrrolidine, rs 1-tertbutoxycarbonyl-3-bromopyrrolidine, rs 1-tert-butoxycarbonyl-3-bromopyrrolidine, 2-methyl-2-propanyl 3-bromo-1-pyrrolidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (+/-)-1-Boc-3-bromopyrrolidine
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only