missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-(bromomethyl)azetidine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 253176-93-1
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD16556174
InChI Key PUKCUGDJEPVLPR-UHFFFAOYSA-N
Synonym 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate
PubChem CID 53350331
IUPAC Name tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CBr)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452160010
SDS View Documents
Thermo Scientific Chemicals
452160010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452160050
SDS View Documents
Thermo Scientific Chemicals
452160050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

253176-93-1
250.14
PUKCUGDJEPVLPR-UHFFFAOYSA-N
53350331
CC(C)(C)OC(=O)N1CC(CBr)C1
C9H16BrNO2
MFCD16556174
1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate
tert-butyl 3-(bromomethyl)azetidine-1-carboxylate

Specifications

253176-93-1
Glass Bottle
MFCD16556174
1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate
CC(C)(C)OC(=O)N1CC(CBr)C1
250.14
250.13
1-BOC-3-(bromomethyl)azetidine
94% min. (GC)
C9H16BrNO2
1g
PUKCUGDJEPVLPR-UHFFFAOYSA-N
tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
53350331
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only