missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-Azetidinone, 97%

Catalog No. AC442040010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442040010 1 g
AC442040050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442040010 Supplier Thermo Scientific Chemicals Supplier No. 442040010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 398489-26-4 | C8H13NO3 | 171.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 398489-26-4
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.20
MDL Number MFCD01861741
InChI Key VMKIXWAFFVLJCK-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinone, 1-boc-azetidin-3-one, 1-n-boc-3-azetidinone, n-boc-3-azetidinone, n-boc-3-oxoazetidine, 1-tert-butoxycarbonyl-3-azetidinone, tert-butyl-3-oxoazetidine-1-carboxylate, 1-boc-3-oxoazetidine, n-boc-azetidin-3-one, 3-oxo-azetidine-1-carboxylic acid tert-butyl ester
PubChem CID 1519404
IUPAC Name tert-butyl 3-oxoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 171.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-BOC-3-Azetidinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only