missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-azetidinemethanol, 95%, Thermo Scientific™

$233.55 - $233.55

Chemical Identifiers

CAS 142253-56-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD06656141
InChI Key HXRDRJKAEYHOBB-UHFFFAOYSA-N
PubChem CID 10583745
IUPAC Name tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CO)C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH57112ME
View Documents
Thermo Scientific Chemicals
H57112ME
500 mg
Each for $233.55
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an intermediate in organic syntheses

Solubility
Soluble in Dimethyl sulfoxide (DMSO).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
Specifications

Chemical Identifiers

142253-56-3
187.24
HXRDRJKAEYHOBB-UHFFFAOYSA-N
tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
C9H17NO3
MFCD06656141
10583745
CC(C)(C)OC(=O)N1CC(CO)C1

Specifications

142253-56-3
C9H17NO3
tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-ylmethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-3-azetidinemethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl3-hydroxymethyl azetidine-1-carboxylate, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester
HXRDRJKAEYHOBB-UHFFFAOYSA-N
1.115
500 mg
117°C (243°F)
187.24
1-Boc-3-azetidinemethanol
268°C to 273°C
MFCD06656141
Soluble in Dimethyl sulfoxide (DMSO).
CC(C)(C)OC(=O)N1CC(CO)C1
tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
187.24
10583745
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents