missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-azetidinemethanol, 95%, Thermo Scientific™

Catalog No. AAH57112ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57112ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57112ME Supplier Thermo Scientific Chemicals Supplier No. H57112ME

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an intermediate in organic syntheses

Solubility
Soluble in Dimethyl sulfoxide (DMSO).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.

Chemical Identifiers

CAS 142253-56-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD06656141
InChI Key HXRDRJKAEYHOBB-UHFFFAOYSA-N
PubChem CID 10583745
IUPAC Name tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CO)C1

Specifications

Boiling Point 268°C to 273°C
Synonym tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-ylmethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-3-azetidinemethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl3-hydroxymethyl azetidine-1-carboxylate, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester
Solubility Information Soluble in Dimethyl sulfoxide (DMSO).
Density 1.115
Quantity 500 mg
Flash Point 117°C (243°F)
Formula Weight 187.24
Percent Purity 95%
Chemical Name or Material 1-Boc-3-azetidinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures