missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-Aminopiperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 436600050

 View more versions of this product

Catalog No. AC436600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-BOC-3-Aminopiperidine
96.0
Authentic
C10H20N2O2
5g
AKQXKEBCONUWCL-UHFFFAOYNA-N
tert-butyl 3-aminopiperidine-1-carboxylate
545809
97%
144243-24-3
100.0
96% min. (GC)
MFCD01861219,MFCD03094717
1-boc-3-aminopiperidine, n-boc-3-aminopiperidine, 3-amino-1-n-boc-piperidine, +/--3-amino-1-n-boc-piperidine, 3-amino-1-boc-piperidine, 1-n-boc-3-aminopiperidine, 3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3-amino-1-tert-butoxycarbonylpiperidine, 3-aminopiperidine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCCC(N)C1
200.28
200.28

Chemical Identifiers

144243-24-3
200.28
AKQXKEBCONUWCL-UHFFFAOYNA-N
545809
CC(C)(C)OC(=O)N1CCCC(N)C1
C10H20N2O2
MFCD01861219,MFCD03094717
1-boc-3-aminopiperidine, n-boc-3-aminopiperidine, 3-amino-1-n-boc-piperidine, +/--3-amino-1-n-boc-piperidine, 3-amino-1-boc-piperidine, 1-n-boc-3-aminopiperidine, 3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3-amino-1-tert-butoxycarbonylpiperidine, 3-aminopiperidine-1-carboxylic acid tert-butyl ester
tert-butyl 3-aminopiperidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only