missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(aminomethyl)azetidine, 97%, Thermo Scientific™

Catalog No. AAH2915203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2915203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2915203 Supplier Thermo Scientific Chemicals Supplier No. H2915203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 325775-44-8
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD01861762
InChI Key XSJPKMUFBHSIRA-UHFFFAOYSA-N
Synonym 1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester
PubChem CID 10679125
IUPAC Name tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)CN

Specifications

Refractive Index 1.467
Quantity 1 g
Formula Weight 186.25
Sensitivity Air Sensitive
Percent Purity 97%
Chemical Name or Material 1-Boc-3-(aminomethyl)azetidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen