missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-aminoazetidine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 193269-78-2
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.23
InChI Key WPGLRFGDZJSQGI-UHFFFAOYSA-N
Synonym 1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
PubChem CID 1516507
IUPAC Name tert-butyl 3-aminoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451180010
SDS View Documents
Thermo Scientific Chemicals
451180010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451180050
View Documents
Thermo Scientific Chemicals
451180050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

193269-78-2
172.23
1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
tert-butyl 3-aminoazetidine-1-carboxylate
C8H16N2O2
WPGLRFGDZJSQGI-UHFFFAOYSA-N
1516507
CC(C)(C)OC(=O)N1CC(C1)N

Specifications

193269-78-2
Glass Bottle
1g
WPGLRFGDZJSQGI-UHFFFAOYSA-N
tert-butyl 3-aminoazetidine-1-carboxylate
1516507
95%
94% min. (GC)
C8H16N2O2
1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
CC(C)(C)OC(=O)N1CC(C1)N
172.23
172.23
1-BOC-3-aminoazetidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only